siRNA Details
Gene Symbol: Spn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009259.5 | NP_033285.1 | 2 | 1314 | ||
GenBank mRNA | AK077764.1 | - | 2 | 1305 | ||
AK134098.1 | - | 2 | 1309 | |||
BC100738.1 | - | 1 | 1206 | 40045521 | ||
BC100739.1 | - | 1 | 1206 | 40045520 | ||
BC100740.1 | - | 1 | 1206 | 40045519 | ||
BC100741.1 | - | 1 | 1206 | 40045517 | ||
S70677.1 | - | 1 | 1206 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
