siRNA Details
Gene Symbol: Prkce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011104.3 | NP_035234.1 | 1 | 1014 | ||
GenBank mRNA | AF028009.1 | - | 1 | 215 | ||
AF325507.1 | - | 1 | 206 | |||
AK042994.1 | - | 1 | 980 | |||
AK082438.1 | - | 1 | 1015 | |||
AK085351.1 | - | 2 | 426 |
