siRNA Details
Gene Symbol: Prkce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011104.3 | NP_035234.1 | 15 | 2959 | ||
GenBank mRNA | AF028009.1 | - | 15 | 2160 | ||
AF325507.1 | - | 15 | 2151 | |||
AK082438.1 | - | 15 | 2962 | |||
AK085351.1 | - | 16 | 2372 |
