siRNA Details
Gene Symbol: PI4KA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058004.3 | NP_477352.3 | 37 | 4446 | ||
XM_005261634.1 | XP_005261691.1 | 36 | 4353 | |||
XM_017028829.1 | XP_016884318.1 | 36 | 4380 | |||
XM_017028830.1 | XP_016884319.1 | 23 | 2602 | |||
GenBank mRNA | AB210002.1 | - | 37 | 4416 | ||
AF012872.1 | - | 36 | 4185 | |||
AK302424.1 | - | 37 | 4374 | |||
AK314781.1 | - | 9 | 1100 | |||
BC018120.1 | - | 7 | 763 | |||
BC053654.1 | - | 6 | 739 | |||
L36151.1 | - | 5 | 732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
