siRNA Details
Gene Symbol: Pou2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011139.2 | NP_035269.2 | 2 | 156 | ||
GenBank mRNA | AK153697.1 | - | 246 | |||
L14677.1 | - | 2 | 154 | |||
Z18537.1 | - | 2 | 157 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pou2f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011139.2 | NP_035269.2 | 2 | 156 | ||
GenBank mRNA | AK153697.1 | - | 246 | |||
L14677.1 | - | 2 | 154 | |||
Z18537.1 | - | 2 | 157 |