siRNA Details
Gene Symbol: Prkg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011160.3 | NP_035290.1 | 1 | 229 | ||
GenBank mRNA | AF084547.1 | - | 1 | 229 | ||
BC113162.1 | - | 1 | 211 | 40060049 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
