siRNA Details
Gene Symbol: Psmb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011187.1 | NP_035317.1 | 7 | 600 | ||
GenBank mRNA | AK002823.1 | - | 7 | 615 | ||
AK012613.1 | - | 7 | 601 | |||
AK013961.1 | - | 7 | 611 | |||
AK031913.1 | - | 7 | 616 | |||
AK075759.1 | - | 7 | 616 | |||
AK088276.1 | - | 7 | 611 | |||
AK088765.1 | - | 7 | 614 | |||
AK168823.1 | - | 7 | 603 | |||
BC033424.1 | - | 7 | 597 | |||
BC049230.1 | - | 7 | 593 | |||
BC057662.1 | - | 7 | 601 | 2865594 | ||
D83585.1 | - | 7 | 600 | |||
Y10874.1 | - | 7 | 614 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
