siRNA Details
Gene Symbol: Rbbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011247.2 | NP_035377.2 | 17 | 2785 | ||
GenBank mRNA | AK144758.1 | - | 2 | 651 | ||
AK161231.1 | - | 2 | 386 | |||
U28789.1 | - | 11 | 1779 | |||
U83913.1 | - | 11 | 1826 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
