siRNA Details
Gene Symbol: Rpl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011291.5 | NP_035421.2 | 5 | 601 | ||
GenBank mRNA | AK017074.1 | - | 5 | 571 | ||
AK050656.1 | - | 5 | 576 | |||
AK088291.1 | - | 5 | 575 | |||
AK135460.1 | - | 4 | 909 | |||
AK146219.1 | - | 5 | 572 | |||
AK150943.1 | - | 5 | 571 | |||
AK167124.1 | - | 5 | 575 | |||
AK168137.1 | - | 5 | 571 | |||
AK168227.1 | - | 5 | 571 | |||
BC025909.1 | - | 5 | 551 | 5027545 | ||
BC051261.1 | - | 5 | 567 | 30072081 | ||
BC086786.1 | - | 4 | 547 | 5701071 | ||
BC096452.1 | - | 5 | 554 | 3588544 | ||
BY441493.1 | - | 1 | 49 | |||
M29016.1 | - | 5 | 570 | |||
M85235.1 | - | 5 | 556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
