siRNA Details
Gene Symbol: Itsn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198968.2 | NP_001185897.1 | 18 | 2273 | ||
NM_011365.4 | NP_035495.2 | 17 | 2192 | |||
GenBank mRNA | AF132479.1 | - | 16 | 1932 | ||
AF132480.1 | - | 16 | 1932 | |||
AK029325.1 | - | 17 | 2139 | |||
AK122480.1 | - | 12 | 1579 | |||
AK161743.1 | - | 6 | 610 | |||
BC138262.1 | - | 18 | 2250 | 8861281 | ||
BC171950.1 | - | 18 | 2250 | 9054236 | ||
BI872589.1 | - | 2 | 160 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK138112.1 | - | 2019 | Chr. - - [-] on Build |
