siRNA Details
Gene Symbol: St6galnac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011372.2 | NP_035502.1 | 2 | 247 | ||
GenBank mRNA | AK021356.1 | - | 1 | 403 | ||
AK034804.1 | - | 2 | 249 | |||
BC058387.1 | - | 2 | 232 | 6817021 | ||
BC131987.1 | - | 2 | 198 | 40130264 | ||
BC132317.1 | - | 2 | 198 | 40130594 | ||
Y11342.1 | - | 2 | 247 |
