siRNA Details
Gene Symbol: St6galnac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011372.2 | NP_035502.1 | 5 | 854 | ||
GenBank mRNA | AK034804.1 | - | 5 | 856 | ||
BC058387.1 | - | 5 | 839 | 6817021 | ||
BC131987.1 | - | 5 | 805 | 40130264 | ||
BC132317.1 | - | 5 | 805 | 40130594 | ||
Y11342.1 | - | 5 | 854 |
