siRNA Details
Gene Symbol: DLGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242761.1 | NP_001229690.1 | 4, 5 | 1492 | ||
NM_001242764.1 | NP_001229693.1 | 1, 2 | 108 | |||
NM_001308390.1 | NP_001295319.1 | 1, 2 | 160 | |||
NM_004746.3 | NP_004737.2 | 4, 5 | 1492 | |||
XM_005258171.2 | XP_005258228.1 | 4, 5 | 1533 | |||
XM_005258172.1 | XP_005258229.1 | 4, 5 | 1528 | |||
XM_011525770.2 | XP_011524072.1 | 4, 5 | 1533 | |||
XM_011525771.2 | XP_011524073.1 | 4, 5 | 1533 | |||
XM_017026085.1 | XP_016881574.1 | 1, 2 | 370 | |||
GenBank mRNA | AB000277.1 | - | 1, 2 | 1032 | ||
AK299880.1 | - | 1, 2 | 108 | |||
AK304101.1 | - | 4, 5 | 1474 | |||
AK316099.1 | - | 4, 5 | 1492 | |||
BC070175.1 | - | 1, 2 | 973 | 6620806 | ||
BC136453.1 | - | 1, 2 | 1024 | 9020442 | ||
BC136454.1 | - | 1, 2 | 1024 | 9020443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
