siRNA Details
Gene Symbol: Spint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011464.2 | NP_035594.1 | 2 | 560 | ||
GenBank mRNA | AF099016.1 | - | 2 | 493 | ||
AI227451.1 | - | 2 | 196 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK171586.1 | - | 1519 | Chr. - - [-] on Build |
