siRNA Details
Gene Symbol: Stk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB015801.1 | - | 10 | 1747 | ||
AF129870.1 | - | 10 | 2125 | |||
AK004499.1 | - | 8 | 1408 | |||
AK172385.1 | - | 10 | 2513 | |||
AK172528.1 | - | 10 | 2510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
