siRNA Details
Gene Symbol: Stx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025307.1 | NP_001020478.1 | 9 | 1369 | ||
NM_001286543.1 | NP_001273472.1 | 10 | 1415 | |||
NM_011502.3 | NP_035632.1 | 9 | 1315 | |||
GenBank mRNA | AK038415.1 | - | 9 | 876 | ||
BC024844.1 | - | 9 | 777 | 5357204 | ||
BC056949.1 | - | 10 | 1415 | |||
D29798.1 | - | 9 | 788 | |||
D29799.1 | - | 9 | 803 | |||
D29800.1 | - | 10 | 747 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
