siRNA Details
Gene Symbol: Scn3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018732.3 | NP_061202.3 | 17 | 3126 | ||
GenBank mRNA | L42337.1 | - | 2 | 1299 |

More Information
Other Species Targeted

Gene Ontology Categories:
