siRNA Details
Gene Symbol: Scn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018733.2 | NP_061203.2 | 27 | 6057 | ||
GenBank mRNA | L42339.1 | - | 10 | 2539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
