siRNA Details
Gene Symbol: Scn9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290674.1 | NP_001277603.1 | 27 | 6522 | ||
NM_001290675.1 | NP_001277604.1 | 27 | 6489 | |||
GenBank mRNA | AK220540.1 | - | 1 | 747 | ||
BC158048.1 | - | 27 | 6180 | 9088161 | ||
BC172147.1 | - | 27 | 6213 | 9054817 | ||
L42338.1 | - | 1 | 961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
