siRNA Details
Gene Symbol: Ptprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029928.2 | NP_084204.2 | 2 | 411 | ||
GenBank mRNA | AK019450.1 | - | 4 | 1074 | ||
AK147439.1 | - | 2 | 413 | |||
AK147668.1 | - | 2 | 413 | |||
AY077755.1 | - | 2 | 289 | |||
BC141006.1 | - | 2 | 207 | 9055618 | ||
BC145111.1 | - | 2 | 207 | 9053659 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008765424, XM_008765425.
Gene Ontology Categories:
