siRNA Details
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177616.1 | NP_001171087.1 | 6 | 690 | ||
NM_001177617.1 | NP_001171088.1 | 6 | 857 | |||
NM_001177618.1 | NP_001171089.1 | 5 | 388 | |||
NM_001177619.1 | NP_001171090.1 | 6 | 1055 | |||
NM_001177623.1 | NP_001171094.1 | 6 | 619 | |||
NM_033264.2 | NP_150289.1 | 5 | 523 | |||
GenBank mRNA | AF324451.1 | - | 5 | 483 | ||
AK029467.1 | - | 6 | 1056 | |||
AK032417.1 | - | 6 | 616 | |||
AK032543.1 | - | 4 | 303 | |||
AK034376.1 | - | 4 | 236 | |||
AK036360.1 | - | 1 | 1353 | |||
AK045507.1 | - | 6 | 1057 | |||
AK050613.1 | - | 6 | 620 | |||
AK137974.1 | - | 6 | 858 | |||
AK153892.1 | - | 7 | 1338 | |||
BC053001.1 | - | 4 | 354 | 5686446 | ||
BC098368.1 | - | 6 | 637 | |||
BC128376.1 | - | 5 | 404 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
