siRNA Details
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177616.1 | NP_001171087.1 | 7 | 820 | ||
NM_001177617.1 | NP_001171088.1 | 7 | 987 | |||
NM_001177618.1 | NP_001171089.1 | 6 | 518 | |||
NM_001177619.1 | NP_001171090.1 | 7 | 1185 | |||
NM_001177623.1 | NP_001171094.1 | 7 | 749 | |||
NM_033264.2 | NP_150289.1 | 6 | 653 | |||
GenBank mRNA | AF324451.1 | - | 6 | 613 | ||
AK029467.1 | - | 7 | 1186 | |||
AK032417.1 | - | 7 | 746 | |||
AK032543.1 | - | 5 | 433 | |||
AK034376.1 | - | 5 | 366 | |||
AK045507.1 | - | 7 | 1187 | |||
AK050613.1 | - | 7 | 750 | |||
AK137974.1 | - | 7 | 988 | |||
AK153892.1 | - | 8 | 1468 | |||
BC053001.1 | - | 5 | 484 | 5686446 | ||
BC098368.1 | - | 7 | 767 | |||
BC128376.1 | - | 6 | 534 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
