siRNA Details
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177616.1 | NP_001171087.1 | 10 | 1143 | ||
NM_001177618.1 | NP_001171089.1 | 9 | 841 | |||
NM_001177619.1 | NP_001171090.1 | 10 | 1508 | |||
NM_033264.2 | NP_150289.1 | 9 | 976 | |||
GenBank mRNA | AF324451.1 | - | 9 | 936 | ||
AK029467.1 | - | 10 | 1509 | |||
AK032417.1 | - | 10 | 1069 | |||
AK032543.1 | - | 8 | 756 | |||
AK034376.1 | - | 8 | 689 | |||
AK153892.1 | - | 11 | 1792 | |||
BC053001.1 | - | 8 | 807 | 5686446 | ||
BC098368.1 | - | 10 | 1090 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001135045, XM_006243973, XM_006243977, XM_008766634, XM_006243982, and others.
Gene Ontology Categories:
