siRNA Details
Gene Symbol: GGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037277.1 | NP_001032354.1 | 2, 3 | 162 | ||
GenBank mRNA | AB016043.1 | - | 2, 3 | 247 | ||
AB017971.1 | - | 1, 2 | 78 | |||
AB019036.1 | - | 2, 3 | 288 | |||
AF057698.1 | - | 2, 3 | 316 | |||
AF125394.1 | - | 2, 3 | 225 | |||
AK025139.1 | - | 2, 3 | 310 | |||
BC005252.1 | - | 2, 3 | 310 | 4051481 | ||
BC067768.1 | - | 2, 3 | 250 | 5295932 | ||
BI462253.1 | - | 2, 3 | 337 | |||
BI520491.1 | - | 2, 3 | 165 | |||
CB216084.1 | - | 2, 3 | 328 | |||
DB473584.1 | - | 2, 3 | 338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
