siRNA Details
Gene Symbol: SH3BP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004844.4 | NP_004835.2 | 2, 3 | 430 | ||
XM_011534251.2 | XP_011532553.1 | 2, 3 | 242 | |||
XM_011534253.1 | XP_011532555.1 | 2, 3 | 391 | |||
XM_017007521.1 | XP_016863010.1 | 2, 3 | 225 | |||
XM_017007522.1 | XP_016863011.1 | 2, 3 | 578 | |||
XM_017007525.1 | XP_016863014.1 | 2, 3 | 92 | |||
GenBank mRNA | AB005047.1 | - | 2, 3 | 182 | ||
AK090524.1 | - | 2, 3 | 173 | |||
AK313167.1 | - | 2, 3 | 388 | |||
AL117422.1 | - | 2, 3 | 387 | |||
BC010123.2 | - | 2, 3 | 361 | 3841593 | ||
BC023195.1 | - | 2, 3 | 102 | |||
BG475051.1 | - | 244 | ||||
BP308415.1 | - | 2, 3 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
