siRNA Details
Gene Symbol: Pde4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011056.3 | NP_035186.1 | 3 | 272 | ||
GenBank mRNA | AF536978.1 | - | 3 | 272 | ||
BC132107.1 | - | 2 | 140 | 40130384 | ||
BC132109.1 | - | 2 | 140 | 40130386 |
