siRNA Details
Gene Symbol: Cdc42ep3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026514.2 | NP_080790.1 | 2 | 1116 | ||
GenBank mRNA | AK012309.1 | - | 2 | 1081 | ||
AK014281.1 | - | 2 | 1118 | |||
AK019447.1 | - | 2 | 1118 | |||
AK150214.1 | - | 2 | 1083 | |||
BC021409.1 | - | 1 | 770 | 5007192 | ||
BC034714.1 | - | 2 | 1069 | 3595315 | ||
BC092297.1 | - | 2 | 1065 | 5063754 |

More Information
Other Species Targeted

Gene Ontology Categories:
