siRNA Details
Gene Symbol: 3110057O12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026622.3 | NP_080898.2 | 1297 | |||
GenBank mRNA | AK028580.1 | - | 1314 | |||
AK031032.1 | - | 1117 | ||||
AK163334.1 | - | 3 | 478 | |||
AK169375.1 | - | 1298 | ||||
BC051081.1 | - | 1367 | ||||
BC092533.1 | - | 2 | 337 | 30503250 | ||
BC145212.1 | - | 804 | 9053763 |
