siRNA Details
Gene Symbol: Nwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177006.3 | NP_795980.2 | 4 | 1583 | ||
GenBank mRNA | AB093290.1 | - | 4 | 452 | ||
AK033855.1 | - | 4 | 1584 | |||
AK034089.1 | - | 5 | 1471 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008770180.
Gene Ontology Categories:
