siRNA Details
Gene Symbol: Nwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177006.3 | NP_795980.2 | 5 | 1750 | ||
GenBank mRNA | AB093290.1 | - | 5 | 619 | ||
AK034089.1 | - | 6 | 1638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
