siRNA Details
Gene Symbol: Tnrc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144925.3 | NP_659174.3 | 22 | 5585 | ||
GenBank mRNA | AK044679.1 | - | 10 | 1969 | ||
AK050601.1 | - | 4 | 465 | |||
AK138426.1 | - | 12 | 1635 | |||
AK142808.1 | - | 7 | 954 | |||
AK142809.1 | - | 7 | 954 | |||
BC005741.1 | - | 11 | 1420 |

More Information
Other Species Targeted

Gene Ontology Categories:
