siRNA Details
Gene Symbol: Tnrc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144925.3 | NP_659174.3 | 24 | 6851 | ||
GenBank mRNA | AK020685.1 | - | 1 | 55 | ||
AK044679.1 | - | 12 | 3235 | |||
AK050601.1 | - | 6 | 1731 | |||
AK138426.1 | - | 14 | 2901 | |||
AK142808.1 | - | 9 | 2220 | |||
AK142809.1 | - | 9 | 2220 | |||
AK147327.1 | - | 24 | 6556 | |||
BC005741.1 | - | 14 | 2348 |

More Information
Other Species Targeted

Gene Ontology Categories:
