siRNA Details
Gene Symbol: Emilin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145158.3 | NP_660140.1 | 4 | 2028 | ||
GenBank mRNA | AF468645.1 | - | 4 | 1979 | ||
AK154230.1 | - | 4 | 1998 | |||
AK155037.1 | - | 4 | 2008 | |||
AK155306.1 | - | 4 | 2022 | |||
AK155366.1 | - | 4 | 2008 | |||
AK155554.1 | - | 4 | 2024 | |||
AK155797.1 | - | 4 | 2029 | |||
AK155878.1 | - | 4 | 2026 | |||
AK155879.1 | - | 4 | 2022 | |||
AK155909.1 | - | 4 | 2028 | |||
AK155913.1 | - | 4 | 1998 | |||
AK155918.1 | - | 4 | 2055 | |||
AK170081.1 | - | 4 | 2025 | |||
AK170288.1 | - | 4 | 1998 | |||
AK170391.1 | - | 4 | 1998 | |||
AK170801.1 | - | 4 | 2029 | |||
AK170822.1 | - | 4 | 2050 | |||
AK170953.1 | - | 4 | 2044 | |||
AK171015.1 | - | 4 | 2050 | |||
AK171020.1 | - | 4 | 2016 | |||
AK171303.1 | - | 4 | 2000 | |||
AK171309.1 | - | 4 | 2025 | |||
AK171579.1 | - | 4 | 1998 | |||
AK171592.1 | - | 4 | 1998 | |||
AK171616.1 | - | 4 | 1998 | |||
BC053753.1 | - | 4 | 2312 | 6510220 | ||
GQ872457.1 | - | 4 | 1923 | |||
GQ872460.1 | - | 4 | 1923 |

More Information
Other Species Targeted

Gene Ontology Categories:
