siRNA Details
Gene Symbol: ZNHIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281433.1 | NP_001268362.1 | 5 | 379 | ||
NM_001281434.1 | NP_001268363.1 | 3 | 245 | |||
NM_004773.3 | NP_004764.1 | 5 | 413 | |||
NR_104009.1 | 5 | 408 | ||||
NR_104010.1 | 4 | 381 | ||||
GenBank mRNA | AB209193.1 | - | 3 | 3877 | ||
AF400652.1 | - | 5 | 405 | |||
AK290858.1 | - | 5 | 364 | |||
BC017931.1 | - | 5 | 378 | 4669658 | ||
BC105608.1 | - | 4 | 353 | |||
BE260293.1 | - | 3 | 144 | |||
BG032116.1 | - | 5 | 314 | |||
BM457955.1 | - | 343 | ||||
CB996852.1 | - | 5 | 439 | |||
HY045632.1 | - | 5 | 414 | |||
L40410.1 | - | 5 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
