siRNA Details
Gene Symbol: B4GALT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004776.3 | NP_004767.1 | 2 | 391 | ||
GenBank mRNA | AB004550.1 | - | 2 | 309 | ||
AB871482.2 | - | 2 | 190 | |||
AF038663.1 | - | 2 | 207 | |||
BC074821.2 | - | 2 | 230 | 30915537 | ||
BC074873.2 | - | 2 | 230 | 30915234 | ||
BC104987.1 | - | 2 | 230 | 8143990 | ||
BC112265.1 | - | 2 | 230 | 8327733 | ||
DR000345.1 | - | 2 | 399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
