siRNA Details
Gene Symbol: Ago2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153178.4 | NP_694818.3 | 19 | 2757 | ||
GenBank mRNA | AB081472.1 | - | 19 | 2619 | ||
AK220193.1 | - | 16 | 2122 | |||
BC023279.1 | - | 11 | 1458 | |||
BC024857.1 | - | 12 | 1602 | |||
BC056639.1 | - | 9 | 1324 | |||
BC064741.1 | - | 14 | 1874 | |||
BC096465.1 | - | 19 | 2662 | |||
BC128379.1 | - | 19 | 2610 | 40111876 | ||
BC129922.1 | - | 19 | 2610 | 40111875 |
