siRNA Details
Gene Symbol: Pkhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153179.3 | NP_694819.2 | 3, 4 | 369 | ||
GenBank mRNA | AK052698.1 | - | 3, 4 | 378 | ||
AK143992.1 | - | 3, 4 | 353 | |||
AY130764.1 | - | 3, 4 | 361 | |||
AY438374.1 | - | 3, 4 | 243 | |||
BB661186.1 | - | 3, 4 | 378 |
