siRNA Details
Gene Symbol: KHK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000221.2 | NP_000212.1 | 1, 2 | 617 | ||
NM_006488.2 | NP_006479.1 | 1, 2 | 617 | |||
XM_005264294.3 | XP_005264351.1 | 1, 2 | 622 | |||
XM_005264296.3 | XP_005264353.1 | 1, 2 | 600 | |||
XM_006712008.3 | XP_006712071.1 | 1, 2 | 622 | |||
XM_006712009.3 | XP_006712072.1 | 1, 2 | 622 | |||
XM_006712010.3 | XP_006712073.1 | 1, 2 | 622 | |||
XM_006712011.3 | XP_006712074.1 | 1, 2 | 622 | |||
XM_006712012.3 | XP_006712075.1 | 1, 2 | 622 | |||
XM_006712013.3 | XP_006712076.1 | 1, 2 | 622 | |||
XM_017004060.1 | XP_016859549.1 | 1, 2 | 622 | |||
XM_017004061.1 | XP_016859550.1 | 1, 2 | 622 | |||
GenBank mRNA | AK130033.1 | - | 1, 2 | 134 | ||
AK223075.1 | - | 1, 2 | 134 | |||
BC006233.2 | - | 1, 2 | 555 | 3946893 | ||
CR456801.1 | - | 1, 2 | 100 | |||
X78678.1 | - | 1, 2 | 108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
