siRNA Details
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172282.2 | NP_758486.1 | 3 | 950 | ||
GenBank mRNA | AK033499.1 | - | 3 | 711 | ||
AK046065.1 | - | 3 | 740 | |||
AK049974.1 | - | 3 | 709 | |||
AK083321.1 | - | 3 | 774 | |||
AK088866.1 | - | 3 | 734 | |||
AK148252.1 | - | 3 | 753 | |||
BC039955.1 | - | 3 | 696 | 4236297 | ||
BC053487.1 | - | 3 | 725 |
