siRNA Details
Gene Symbol: Tmco3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172282.2 | NP_758486.1 | 3, 4 | 1066 | ||
GenBank mRNA | AK033499.1 | - | 3, 4 | 827 | ||
AK046065.1 | - | 3, 4 | 856 | |||
AK049974.1 | - | 3, 4 | 825 | |||
AK083321.1 | - | 3, 4 | 890 | |||
AK088866.1 | - | 3, 4 | 850 | |||
AK148252.1 | - | 3, 4 | 869 | |||
BC039955.1 | - | 3, 4 | 812 | 4236297 | ||
BC053487.1 | - | 3, 4 | 841 |
