siRNA Details
Gene Symbol: Plcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172285.1 | NP_758489.1 | 20, 21 | 2426 | ||
GenBank mRNA | AK142173.1 | - | 20, 21 | 2429 | ||
AK170430.1 | - | 20, 21 | 2435 | |||
BC019654.1 | - | 7, 8 | 960 | |||
BC023877.1 | - | 20, 21 | 2426 | 5354925 | ||
BC037700.1 | - | 2, 3 | 218 | |||
BC049273.1 | - | 3, 4 | 352 |
