siRNA Details
Gene Symbol: Sorbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205219.1 | NP_001192148.1 | 24 | 5181 | ||
NM_172752.4 | NP_766340.4 | 23 | 5127 | |||
GenBank mRNA | AK041051.1 | - | 4 | 1414 | ||
AK122369.1 | - | 19 | 4592 | |||
AK135952.1 | - | 2 | 1302 | |||
AK140498.1 | - | 19 | 3366 | |||
BC031203.1 | - | 7 | 1900 | |||
BC039163.1 | - | 23 | 3501 | |||
BC151038.1 | - | 24 | 5051 | 9087951 | ||
BC157944.1 | - | 20 | 3272 | 9007279 | ||
BC172140.1 | - | 23 | 4997 | 9054810 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
