siRNA Details
Gene Symbol: Zfp809 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164624.1 | NP_001158096.1 | 5 | 1319 | ||
GenBank mRNA | AK044107.1 | - | 3 | 1115 | ||
AK049344.1 | - | 5 | 1280 | |||
AK089532.1 | - | 5 | 1483 | |||
BY764604.1 | - | 1 | 422 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
