siRNA Details
Gene Symbol: BMPR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204.6 | NP_001195.2 | 4, 5 | 1685 | ||
XM_011511687.1 | XP_011509989.1 | 4, 5 | 1702 | |||
GenBank mRNA | AK292430.1 | - | 4, 5 | 949 | ||
BC052985.2 | - | 4, 5 | 1111 | 6149698 | ||
D50516.1 | - | 4, 5 | 945 | |||
U20165.1 | - | 4, 5 | 573 | |||
Z48923.1 | - | 4, 5 | 918 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
