siRNA Details
Gene Symbol: Sowahc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172939.3 | NP_766527.3 | 1 | 1156 | ||
GenBank mRNA | AK030944.1 | - | 1 | 1157 | ||
AK050497.1 | - | 1 | 1127 |

More Information
Other Species Targeted

Gene Ontology Categories:
