siRNA Details
Gene Symbol: Rnf213 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077880.1 | - | 5, 6 | 707 | ||
AK156571.1 | - | 6, 7 | 840 | |||
AK173199.1 | - | 15, 16 | 4053 | |||
BC038025.1 | - | 7, 8 | 998 | |||
BC119284.1 | - | 4, 5 | 528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
