siRNA Details
Gene Symbol: Myo9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173018.2 | NP_766606.2 | 33, 34 | 6437 | ||
GenBank mRNA | AK028873.1 | - | 4, 5 | 440 | ||
AK029836.1 | - | 3, 4 | 304 | |||
BC068298.1 | - | 7, 8 | 875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
