siRNA Details
Gene Symbol: Myo9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173018.2 | NP_766606.2 | 44 | 9125 | ||
GenBank mRNA | AK028873.1 | - | 14 | 3074 | ||
AK029836.1 | - | 13 | 2938 | |||
AK162486.1 | - | 1 | 1252 | |||
BC046526.1 | - | 7 | 2187 | |||
BC068298.1 | - | 17 | 3506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
