siRNA Details
Gene Symbol: MAGI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004742.2 | NP_004733.2 | 23 | 4534 | ||
GenBank mRNA | AB010894.1 | - | 23 | 4250 | ||
AK289803.1 | - | 23 | 4103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
