siRNA Details
Gene Symbol: Nbeal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173444.2 | NP_775620.2 | 45 | 7135 | ||
GenBank mRNA | AK041108.1 | - | 5 | 473 | ||
BC055813.1 | - | 8 | 864 | |||
BC060141.1 | - | 21 | 3266 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
